missing translation for 'onlineSavingsMsg'
Learn More

Elaidic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012505
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012505 5 g
111012504 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012505 Supplier Merck Emd Millipore Supplier No. 450895G
Only null left

Specifications

Packaging Poly Bottle
CAS 112-79-8
Flash Point >113°C
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COOH
MDL Number MFCD00063954
Melting Point 42°C to 44°C
Boiling Point 288°C (100 mmHg)
Synonym trans-9-Octadecenoic acid; trans-Oleic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 282.46
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less