missing translation for 'onlineSavingsMsg'
Learn More

Elaidic acid, 98%

Catalog No. AAA1483214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1483214 25 g
AAA1483203 1 g
AAA1483206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1483214 Supplier Thermo Scientific Chemicals Supplier No. A1483214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Store cold | CAS: 112-79-8 | C18H34O2 | 282.468 g/mol

In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 112-79-8
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00063954
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
PubChem CID 637517
ChEBI CHEBI:27997
IUPAC Name (E)-octadec-9-enoic acid
SMILES CCCCCCCCC=CCCCCCCCC(=O)O

Specifications

Melting Point 41°C to 45°C
Boiling Point 288°C (100 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 98%
Quantity 25 g
Beilstein 1726543
Merck Index 14,3533
Solubility Information Insoluble in water.
Formula Weight 282.47
Percent Purity 98%
Chemical Name or Material Elaidic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-006-6
RTECSNumber JX6125000
TSCA No
storageNote1 Air sensitive
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only