Learn More
Eisco™ Water Turbine with Dynamo/Hydroelectrical Station
Catalog No. S05837
Change view
Click to view available options
Product Type:
Hydro-Electric Power Station
Catalog No. Product Type
S05837 Hydro-Electric Power Station
1 options
Catalog No. S05837 Supplier Eisco™ Supplier No. PH0497AN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Convert mechanical energy into electrical energy.
  • Consists of transparent cover, inlet and outlet tubes directly connected to a dynamo
  • Fitted on base
Specifications
Product Type Hydro-Electric Power Station
Size Length: 13.77 in.
For Use With (Application) Convert mechanical energy into electrical energy
Includes Transparent cover, inlet and outlet tubes directly connected to a dynamo
Weight (English) 4.34 lb.
Weight (Metric) 1.97 kg