missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Pulley Bench Clamp Fitting

Eisco bench clamp pulley with metal ball bearing carried on a two part aluminum frame is designed primarily for clamping to the edge of a bench, table etc. when used in classroom science experiments.

Supplier:  Eisco™ PH0296B

 Afficher plus de versions de ce produit

Numéro de catalogue. S99356


Il en reste null

Description

Description

  • 5.0cm diameter pulley carried on two-part aluminum frame
  • Use to clamp to the edge of a bench or table
  • Pulley overhangs the edge in a vertical plane

Order Info

Overall length of frame (pulley bar): 4.3 in. (11.0cm); maximum clamp opening: 1 in. (2.5cm).

Spécifications

Identifiants chimiques

Pulley Bench Clamp Fitting
For clamping to the edge of a bench, table
12 cm
1.96 in.
5 cm
4.72 in.

Spécifications

Pulley Bench Clamp Fitting
1.96 in.
5 cm
4 cm
5.51 in.
0.21 kg
12 cm
14 x 12 x 4 cm
Aluminum, Nylon
For clamping to the edge of a bench, table
1.57 in.
Two part Aluminum frame, pulley bar frame, Nylon ball bearing pulley
14 cm
0.46 lb.
4.72 in.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions