missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Double Burette Clamp

Catalog No. S07858
Change view
Click to view available options
Jaw Type:
Spring Loaded
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Jaw Type
S07858 Spring Loaded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07858 Supplier Eisco™ Supplier No. CH0258B
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suited to hold two burettes up to 100mL.

  • Metal construction for high strength
  • With jaws and rubber projected tips

Specifications

Type Burette Clamp (Double)
Material High Strength Alloy
For Use With (Equipment) Burette
Length (English) 11.41 in.
Length (Metric) 29 cm
Color Blue
Jaw Type Spring Loaded