Learn More
Eisco™ Direct Vision Spectroscope in Metal Case
Catalog No. S12964
Change view
Click to view available options
Product Type:
Direct Vision Spectroscope
Catalog No. Product Type
S12964 Direct Vision Spectroscope
1 options
Catalog No. S12964 Supplier Eisco™ Supplier No. PH0595
Includes
Plastic vials with a mount onto the spectroscope.
Sturdy housing is ideal for student use.
  • Includes a very bright 600 lines/mm holographic replica granting a very good resolution
  • Bright bands of the dominant wavelengths are clearly visible against the scale
  • Printed scale in english on metal case
  • Case Dimensions: 7 x 4.75 x 1.12 in.
Specifications
For Use With (Application) Lets users look through a split spectrum of light decomposed by an optical system.
Includes Plastic vials with a mount onto the spectroscope.
Product Type Direct Vision Spectroscope