missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4H Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan
WBSCR1 is one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization.This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
Spécifications
Spécifications
| Antigène | eIF4H |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | EIF4H |
| Numéro d’acquisition du gène | Q15056 |
| Alias du gène | Williams-Beuren syndrome chromosome region 1; eIF-4H; williams-Beuren syndrome chromosomal region 1 protein |
| Symboles de gène | EIF4H |
| Afficher plus de résultats |