missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EIF2AK2 (PKR), GST Tag Recombinant Protein

Numéro de catalogue. PV4821
Encompass
Modifier l'affichage
Click to view available options
Quantité:
10 μg
100 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PV4821 10 μg
PR8917B 100 μg
PV4822 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PV4821 Fournisseur Invitrogen™ Code fournisseur PV4821
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

EIF2AK2 (PKR) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses. EIF2AK2 plays a key role in antiviral defense mediated through its direct activation by double-stranded RNA produced by viral infection. PKR plays a key role in IFN induced-innate antiviral response, virus-induced apoptosis, cell growth & differentiation. PKR induces apoptosis by up-regulating Fas expression and mediates FADD/Caspase-8 death signaling pathway. Upon viral infection, it functions as a dual protein, sequentially activating both cell survival & cell death pathways using kinase independent and dependent strategies. PRKR regulates multiple pathways which include NF-kappaB activation, p53, p38, & PDGF signaling pathway. PKR has been implicated in tumor suppression & malignancy. To evade the antiviral effects of PKR, viruses have evolved multiple mechanisms, such as the inhibition of PKR by the non-structural protein (NS1) of the influenza virus. More recently, PKR has been implicated in several neurodegenerative diseases including Alzheimer, Huntington, and amyotrophic lateral sclerosis.

Spécifications

Numéro d'enregistrement P19525
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM tris with 2mM DTT, 50% glycerol, 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5
ID du gène (Entrez) 5610
Poids moléculaire (g/mol) 62.2 kDa
Nom Human EIF2AK2 (PKR), GST Tag
Plage de pH 7.5
Méthode de purification Purified
Quantité 10 μg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
Statut réglementaire RUO
Alias du gène 2310047A08Rik; 4732414G15Rik; AI467567; AI747578; double stranded RNA activated protein kinase; dsRNA-activated kinase; EC 2.7.11.1; eIF-2 alpha; eIF2a Kinase; eIF-2A protein kinase 2; EIF2AK1; EIF2AK2; eukaryotic translation initiation factor 2 alpha kinase 2; eukaryotic translation initiation factor 2-alpha kinase 2; IFN- type I-induced and dsRNA-activated kinase; IFN-induced and double-stranded RNA-activated kinase; interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; interferon-inducible RNA-dependent protein kinase; MGC126524; OTTHUMP00000201320; P1/eIF-2A protein kinase; p68 kinase; PKR; PPP1R83; Prkr; protein kinase R; protein kinase RNA-activated; protein kinase, interferon inducible double stranded RNA dependent; Protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; serine/threonine-protein kinase TIK; T-cell viral integration site; Tik; Tyrosine-protein kinase EIF2AK2
Nom usuel PKR
Symbole de gène EIF2AK2
KinaseFamily PEK Kinase Family
Longueur de protéine 252-551
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Étiquette de la protéine GST-tag
Séquence 252-551
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Saisie d’essai Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Groupe kinase Ser⁄Thr Kinases: Other
Shipping condition (Condition d'expédition) Dry Ice
Compatibilité du système LanthaScreen™ Antibody, LanthaScreen™ Substrate
Saisie de la cible EIF2AK2
Gène cible eukaryotic translation initiation factor 2-alpha kinase 2
Système d’expression Insect cells
Famille de protéines Kinases & Inhibitors
Forme de protéine Recombinant, Catalytic
Forme Liquid
Sous-type de protéine Serine/Threonine Kinases
Catégorie de recherche Signal Transduction
Afficher plus de résultats Afficher moins de résultats