Learn More
Abcam Inc EI1, EZH2 inhibitor, 50MG
Numéro de catalogue. 502672012
Encompass
MW 390.5 Da. Potent (IC = 15 nM for wild-type enzyme; 13 nM for Y641F mutant enzyme) EZH2 inhibitor. Selective versus EZH1 (142-fold). Highly selective (>10,000-fold) against a panel of 10 other human methyltransferases. Inhibits cellular H3K27 methylation and activates Ezh2 target gene p16 expression in diffuse large B-cell lymphoma cells (DLBLC). Selectively inhibits growth and promotes cell cycle arrest and apoptosis of DLBLC cells with Y641 EZH2 mutations.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502672012 Fournisseur Abcam Inc Code fournisseur AB26981750MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 390.5 Da. Potent (IC = 15 nM for wild-type enzyme; 13 nM for Y641F mutant enzyme) EZH2 inhibitor. Selective versus EZH1 (142-fold). Highly selective (>10,000-fold) against a panel of 10 other human methyltransferases. Inhibits cellular H3K27 methylation and activates Ezh2 target gene p16 expression in diffuse large B-cell lymphoma cells (DLBLC). Selectively inhibits growth and promotes cell cycle arrest and apoptosis of DLBLC cells with Y641 EZH2 mutations.
The product is subject to the following: Abcam Restricted Use Statement