Learn More
SignalChem Biotech CDK1/CyclinE1, Active 50ug
Numéro de catalogue. 502575651
Encompass
Recombinant full-length human CDK1 and Cyclin E1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
Numéro de catalogue. 502575651 Fournisseur SignalChem Biotech Code fournisseur C2218CG50
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant full-length human CDK1 and Cyclin E1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.