missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEA1 Recombinant Rabbit Monoclonal Antibody (23GB5260)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553877
Description
EEA1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Early endosomal antigen 1 (EEA1) is a 162 kDa membrane bound protein component specific to the early endosome and is essential for fusion between early endocytic vesicles. Early endosomes are cytoplasmic compartments which fuse with endocytic vesicles for redistribution of extracellular compounds to alternate destinations. Zinc-finger-like domains, reminiscent of those found in nucleic acid binding proteins, are located in the amino and carboxyl-terminal domains of EEA1. The carboxyl-terminal zinc-finger-like-domain is conserved in several other non-nuclear proteins, some of which are also involved in intracellular protein trafficking. In addition, this domain is an authentic zinc-binding domain and is critical to the intracellular localization of EEA1. Membrane association of EEA1 has been shown dependent on phosphatidyl 3-kinase activity, inhibitors of which cause EEA1 to dissociate from early endosomes.Identifiants chimiques
| EEA1 | |
| 1.5 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15075, Q8BL66 | |
| 216238, 314764, 8411 | |
| A synthesized peptide derived from human EEA1 (1-63AA). | |
| Antibody |
| 23GB5260 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| EEA1 | |
| A430109M19Rik; B230358H09Rik; Early endosome antigen 1; early endosome antigen 1, 162kD; early endosome-associated protein; EEA1; Endosome-associated protein p162; MST105; MSTP105; ZFYVE2; Zinc finger FYVE domain-containing protein 2 | |
| EEA1 | |
| Primary | |
| Affinity chromatography |
Spécifications
| EEA1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15075, Q8BL66 | |
| EEA1 | |
| A synthesized peptide derived from human EEA1 (1-63AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5260 | |
| Unconjugated | |
| EEA1 | |
| A430109M19Rik; B230358H09Rik; Early endosome antigen 1; early endosome antigen 1, 162kD; early endosome-associated protein; EEA1; Endosome-associated protein p162; MST105; MSTP105; ZFYVE2; Zinc finger FYVE domain-containing protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216238, 314764, 8411 | |
| -20°C | |
| Liquid |