missing translation for 'onlineSavingsMsg'
Learn More

Edvotek™ Sickle Cell Gene Detection

Catalog No. S94670
Encompass
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94670 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94670 Supplier Edvotek™ Supplier No. 116

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets

Requires

Horizontal gel electrophoresis apparatus; D.C. power supply; Automatic micropipets with tips; Balance; Microwave, hotplate or burner; Pipet pump; 250mL flasks or beakers; Hot gloves; Safety goggles and disposable laboratory gloves; Small plastic trays or large weigh boats (for gel destaining); DNA visualization system (white light); Distilled or deionized water

Gain an understanding of the effect of mutations on health and disease.

  • Genetically inherited diseases in humans can be caused by a single nucleotide mutation resulting in a change. Such a substitution can affect the structure and function of a critical protein, rendering it partially or completely nonfunctional. A classic example is hemoglobin in sickle cell anemia
  • Agarose gel-based experiment designed for DNA staining and visualization with InstaStain™ Methylene Blue
  • Demonstrates the use of a restriction enzyme that discriminates between the normal and sickle cell hemoglobin genes
  • This experiment does not contain human DNA

Specifications

Grade 9 to Undergraduate
For Use With (Application) To investigate the restriction enzyme that discriminates between hba (normal) and hbs (disease) genes and perform a simulated test on a patient
Includes Instructions; ready-to-load™ quickstrip™ dna samples; ultraspec agarose™ powder; practice gel loading solution; electrophoresis buffer; instastain™ blue and flashblue™
Quantity 8 Gels
Requires Electrophoresis apparatus, power supply, automatic micropipet and tips, balance, microwave or hot plate, visualization system
Time Range 45 min.
Content And Storage Refrigerated
Class Size 8 Lab Groups
Product Type Sickle-cell Gene Detection (DNA-based) Classroom Kit