missing translation for 'onlineSavingsMsg'
Learn More

Edvotek™ DNA Fingerprinting by PCR Amplification

Catalog No. S94607
Encompass
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94607 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94607 Supplier Edvotek™ Supplier No. EDV130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets

Requires

Horizontal gel electrophoresis apparatus; DC power supply; Automatic micropipets with tips; Balance; Microwave, hot plate or burner; Pipet pump; 250mL flasks or beakers; Hot gloves; Safety goggles and disposable laboratory gloves; Small plastic trays or large weigh boats (for gel destaining); DNA visualization system (white light); Distilled or deionized water

Demonstrates the utility of DNA amplification in forensics.

  • Forensic DNA fingerprinting has become a universally accepted crime-fighting tool. Recent advances use the Polymerase Chain Reaction (PCR) to amplify human DNA obtained from crime scenes
  • Develop a basic understanding of DNA fingerprinting
  • Experiment is based on a crime scene scenario and has an inquiry-based component
  • Enables varied results depending on the selection of DNA fingerprinting patterns
  • Students analyze PCR reactions obtained from different suspects and compare them to a crime scene sample
  • Does not contain human DNA

Specifications

Grade 9 to Undergraduate
For Use With (Application) For DNA fingerprinting
Quantity 8 Gels
Requires Electrophoresis apparatus; power supply; automatic micropipet and tips; balance; microwave or hot plate; visualization system
Time Range 45 min.
Content And Storage Refrigerated
Class Size 8 Lab Groups
Product Type DNA Fingerprinting By PCR Amplification Classroom Kit