missing translation for 'onlineSavingsMsg'
Learn More

Edvotek™ Analysis of Restriction Enzyme Cleavage Patterns of DNA

Catalog No. S11102
Encompass
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11102 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11102 Supplier Edvotek™ Supplier No. EDV102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets.

Requires

Horizontal electrophoresis apparatus; DC power supply; Staining tray and net; Visualization system (white light); Automatic micropipets with tips; 250mL flasks or beakers; Distilled or deionized water; Hotplate, Bunsen burner or microwave; Balance

Observe patterns and frequency of cleavage through agarose gel electrophoresis.

Plasmid and lambda DNA are pre-digested with restriction enzymes - endonucleases that recognize and cut double-stranded DNA within or near defined base sequences. Digests are separated by agarose gel electrophoresis.

Specifications

Grade 9 to Undergraduate
Quantity 8 Gels
Class Size 8 Lab Groups
Product Type Electrophoresis Kit