missing translation for 'onlineSavingsMsg'
Learn More

EDTA Stabilizer, 50% (w/v) EDTA and 10% (w/v) NaOH, Ricca Chemical

Catalog No. 268032
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
268032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 268032 Supplier Ricca Chemical Company Supplier No. 268032
Only null left

For Ammonia Nitrogen Determination

Chemical Identifiers

CAS 6381-92-6 , 7732-18-5 , 1310-73-2
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 131664146
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless to Yellow
CAS Min % 30.64, 61.0, 6.13
CAS Max % 31.89, 64.0, 6.38
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼9.5
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 50% (w/v) EDTA + 10% (w/v) NaOH
Chemical Name or Material EDTA Stabilizer
Show More Show Less