missing translation for 'onlineSavingsMsg'
Learn More

EDTA Reagent, For Nitrogen (Ammonia), Nitrogen (Organic), 5% (w/v), LabChem™

$91.25 - $138.00

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 131664146
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC139301
View Documents
LabChem
LC139301
500 mL
Each for $91.25
Only null left
 
LC139302
View Documents
LabChem
LC139302
1 L
Each for $138.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
131664146
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate

Specifications

Colorless
5,94.95,0.05
(HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
131664146
5% (w/v)
Poly Bottle
500 mL
EDTA Reagent, For Nitrogen (Ammonia), Nitrogen (Organic)
6381-92-6 , 7732-18-5 , 1310-73-2
C10H18N2Na2O10
MFCD00150037,MFCD00003541
Soluble in water
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.23
Passes Test
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents