missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Disodium, 0.01M, 1 mL = 1 mg CaCO3 With Mg, Solution, Spectrum™ Chemical
$868.66 - $868.66
Specifications
| Chemical Name or Material | EDTA Disodium |
|---|---|
| CAS | 14402-88-1 , 7732-18-5 , 6381-92-6 |
| Concentration | 0.01 M ± 0.0002 M |
| Grade | Chemical Solution |
| Packaging | Poly Bottle |
Specifications
| EDTA Disodium | |
| 0.01 M ± 0.0002 M | |
| Poly Bottle |
| 14402-88-1 , 7732-18-5 , 6381-92-6 | |
| Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.