missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium, 0.01M, 1 mL = 1 mg CaCO3 With Mg, Solution, Spectrum™ Chemical

Catalog No. 18610326
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610326 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18610326 Supplier Spectrum Chemical Mfg Cor Supplier No. E1101LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material EDTA Disodium
CAS 14402-88-1, 7732-18-5, 6381-92-6
Concentration 0.01 M ± 0.0002 M
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.