missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetate Disodium U.S.P., Macron Fine Chemicals™
Specifications
Specifications
| Packaging | POLYSTORMOR |
| Quantity | 500 g |
| IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
| Formula Weight | 372.24 |
| Percent Purity | 99.0 to 101.0% |
| Grade | USP |
| Chemical Name or Material | Edetate Disodium |