missing translation for 'onlineSavingsMsg'
Learn More

Edetate Disodium, Dihydrate, BiotechGrade, 99-101%, Spectrum™ Chemical

Catalog No. 18602384
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602384 45 kg Fiber Drum
18602379 500 g Poly Bottle
18602381 2.5 kg Poly Bottle
18602380 6 x 500 g Poly Bottle
18602382 4 x 2.5 kg Poly Bottle
18602383 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602384 Supplier Spectrum Chemical Mfg Cor Supplier No. E105845KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E1058, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical Edetate Disodium, Dihydrate, BiotechGrade is an aminopolycarboxylic acid and a colorless, water-soluble solid. It is useful in the laboratory as a hexadentate ligand and chelating agent. The GR brand is an EMD Chemical trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 8.7-11.4%
Packaging Fiber Drum
Quantity 45 kg
Percent Purity 99 to 101%
Grade Biotech