missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDB/DBCP - WP (Low Level), Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C - closed cup |
| Type | Certified Reference Material |
| Quantity | 1.2 mL |
| Grade | Certified Reference Material |