missing translation for 'onlineSavingsMsg'
Learn More

Mopec Economy Hospital Body Bag with Envelope Zipper

Catalog No. BE082
Change view
Click to view available options
Dimensions (L x W):
90 x 36in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
BE082 90 x 36in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BE082 Supplier Mopec Supplier No. BE082

Mopec Economy Hospital Body Bag with Envelope Zipper features a rust resistant nylon zipper with two pulls

  • No handles
  • Shelf life:10 years

Specifications

Closure Type Zip Closure
Color White
Material Virgin PVC
Quantity 1
Dimensions (L x W) 90 x 36in.