missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Economy Electricity Kit

Catalog No. S23691
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S23691 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23691 Supplier Eisco™ Supplier No. PH1057

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for introductory electronics. The Economy Electricity Kit is a basic assortment of circuit components.

  • Ideal for introductory electronics.

Specifications

Grade 6 to 12
Includes 10 MES Lamp Holders, 10 MES Bulbs, (1) 6 V Buzzer with Connecting Leads, (1) Electric Motor, (3) 6 V, 15 Slide Switches, (1) 1.5 V Cell Holder, (10) Crocodile Leads
Class Size 1 Lab Group
Product Type Economy Electricity Kit