missing translation for 'onlineSavingsMsg'
Learn More

Econazole Nitrate 98.0+%, TCI America™

Catalog No. E09575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09575G 5 g
E095725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09575G Supplier TCI America Supplier No. E09575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24169-02-6
Molecular Formula C18H16Cl3N3O4
Molecular Weight (g/mol) 444.693
MDL Number MFCD00058160
InChI Key DDXORDQKGIZAME-UHFFFAOYSA-N
Synonym econazole nitrate, ifenec, epi-pevaryl, gyno-pevaryl, econazole nitrate salt, pevaryl, +--econazole nitrate, spectazole, econazolum nitrate, gyno-pevaryl 150
PubChem CID 68589
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
SMILES C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]

Specifications

Melting Point 161°C
Color White
Quantity 5 g
Formula Weight 444.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Econazole Nitrate
RTECSNumber NI4450000
TSCA No