Learn More
Lonza Walkersville Inc EBM-2 Endothelial Cell Growth Basal Medium-2, 500 mL
Numéro de catalogue. NC1447083
Encompass
EBM -2 is an endothelial basal medium sold separately. This can be used with 1 x kit of CC-4176 (Large-vessel endothelial cell growth medium SingleQuots Supplements and Growth Factors) or 1 x CC-4147 (Microvascular endothelial cell growth medium SingleQuots Supplements and Growth Factors).Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed.Content:1 x Bottle EBM-2 Basal Medium, 500 mL
Numéro de catalogue. NC1447083 Fournisseur Lonza Walkersville Inc Code fournisseur CC3156
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
EBM -2 is an endothelial basal medium sold separately. This can be used with 1 x kit of CC-4176 (Large-vessel endothelial cell growth medium SingleQuots Supplements and Growth Factors) or 1 x CC-4147 (Microvascular endothelial cell growth medium SingleQuots Supplements and Growth Factors).Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed.Content:1 x Bottle EBM-2 Basal Medium, 500 mL