Learn More
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
Catalog No. 501129264
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129264 100 μL
501129756 4 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501129264 Supplier Invitrogen™ Supplier No. 00497793
Only null left
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer.
  • A ready to use solution of PHA-L in aqueous buffer
  • PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris
  • It is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity
  • This reagent is intended for use in in vitro activation of human and mouse leukocytes
Specifications
Quantity 100 μL
Form Liquid
Product Line eBioscience
Product Type Phytohemagglutinin-L (PHA-L) Solution
For Research Use Only. Not for use in diagnostic procedures.