missing translation for 'onlineSavingsMsg'
Learn More

EarthBox™ Container Garden System

Catalog No. S93903
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S93903 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S93903 Supplier EarthBox™ Supplier No. EDURTGGRNPALL9

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • EarthBox
  • Aeration screen
  • Water fill tube
  • Two fitted mulch covers
  • Instruction sheet and planting guide

Requires

Seeds or seedlings, proper lighting and water

Container Garden System makes it easy for you to start your very own container garden.

This portable, easy-to-use scientifically engineered garden system requires no weeding, hoeing or digging. Advances the study of the science behind plants, water, soil, light and nutrition while promoting food security, conservation and the sustainability of nonrenewable resources.

Specifications

Color Green
For Use With (Application) Growing a garden in or outside a classroom
Includes Earthbox™ container, aeration screen, water fill tube, 2 fitted mulch covers, instruction sheet and planting guide
Quantity 1 Kit
Requires Seeds or seedlings, proper lighting and water
Dimensions (L x W x H) L: 29 in.; H: 14 in.; D: 11 in.
Product Type Earthbox™ Container Garden Kit