Learn More
Invitrogen™ E-Gel™ 1 Kb Plus Express DNA Ladder
Description
Includes
DNA Ladder and 1 vial 1X E-Gel™ Sample Loading Buffer
Invitrogen E-Gel 1 Kb Plus Express DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 5,000 bp. E-Gel 1 Kb Plus Express DNA Ladder consists of nine individual chromatography-purified DNA fragments with reference bands at 1,500 and 500 bp for easy orientation.
E-Gel 1 Kb Plus Express DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.
Highlights of E-Gel 1 Kb Plus Express DNA Ladder:
- Performance—optimized for use on E-Gel CloneWell ll agarose gels
- Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature up to six months
- Ready to use—premixed with loading buffer
- Convenient—provided with 1X E-Gel Sample Loading Buffer
- Precise—an exact amount of DNA in each band
Specifications
Specifications
| Content And Storage | • 2 x 1.25 mL E-Gel 1 Kb Plus Express DNA Ladder • 1.25 mL 1X E-Gel Sample Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods store at -20°C. |
| Concentration | 0.04 μg/μL |
| Gel Type | Agarose |
| Ready to Load | Yes |
| Sample Loading Volume | 1.25 mL |
| Size Range | 100 to 5000 bp |
| Volume (Metric) | 2 x 1.25 mL |
| Gel Compatibility | CloneWell™ II E-Gels, Single Comb E-Gels |
| Quantity | 100 applications |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
| Show More |
Frequently Asked Questions (FAQs)
Sequences of Invitrogen DNA and RNA ladders are proprietary.
Invitrogen DNA ladders contain linear dsDNA fragments.
Invitrogen DNA ladders are composed of double-stranded DNA fragments only.
This can happen if the marker was heated. Please ensure that the ladders are not heated before use.
TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.
TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.
The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.
Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.
For Research Use Only. Not for use in diagnostic procedures.