missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-Methylglutaconic acid, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014070
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014070 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014070 Supplier Merck Emd Millipore Supplier No. 4410850MG

Chemical Identifiers

MDL Number MFCD01556044
Synonym (2 E)-3-Methyl-2-pentenedioic acid

Specifications

Melting Point 137°C to 143°C
CAS 372-42-9
Linear Formula C6H8O4
Quantity 50 mg
Formula Weight 144.13
Percent Purity ≥97.0% (HPLC)