missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLDY22X

 View more versions of this product

Catalog No. PLDY2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Dy1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Dysprosium Standard
7429-91-6 , 7697-37-2 , 7732-18-5
0.1,2.0,97.9
Dy
AA, ICP
Certified Reference Material
[Dy]
dysprosium
500 mL
CHEBI:33377
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLDY2-2Y, -2X    10,000    PLDY2-3Y, -3X
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00010982
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
KBQHZAAAGSGFKK-UHFFFAOYSA-N
2% HNO3
162.50
23912
Assurance

Chemical Identifiers

7429-91-6
162.50
KBQHZAAAGSGFKK-UHFFFAOYSA-N
CHEBI:33377
[Dy]
Dy
MFCD00010982
23912
dysprosium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only