missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta)

Catalog No. AA40285HF
Change view
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40285HF 10 mm
AA40285DH 25 mm
AA40285DJ 50 mm
AA40285DM 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA40285HF Supplier Thermo Scientific Chemicals Supplier No. 040285HF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx. 0.27g/mm, Cast rod, diameter is approximate | CAS: 7429-91-6 | Dy | 162.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Gray to Silver
Physical Form Cast Rod
Quantity 10 mm
Assay Percent Range 99.7% (Metals basis excluding Ta)
Odor Odorless
Appearance Gray to silver rod
Chemical Name or Material Dysprosium
EINECSNumber 231-073-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only