missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Dynamic Weighing Kit

Catalog No. 01919109
Change view
Click to view available options
0.001 g/0.01 g Platform
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01919109 0.001 g/0.01 g Platform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01919109 Supplier Mettler Toledo™ Supplier No. 11132657

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Small, 1 and 2 Place Weighing Pans on Precision Balances (XPR/XSR/XPE/XP/XS)

  • Dynamic Weighing Kit
  • Contaminant Protection
  • Easy to use
  • For dynamic weighing with 4 L vessel
  • For small, 1 and 2 place weighing pans
  • 0.001 g/0.01 g Platform
  • Compatible with XPR/XSR/XPE/XP/XS Precision Balances
  • Start to measure dynamically after quick and simple installation

Specifications

For Use With (Equipment) XPR/XSR/XPE/XP/XS Precision Balances
For Use With (Application) Dynamic Weighing
Size 0.001 g/0.01 g Platform
Type Balance Accessories
Autoclavable Not Autoclavable