missing translation for 'onlineSavingsMsg'
Learn More

Dynamic Diagnostics Monarch™ 1153™ Three-Line Label Gun

Catalog No. 22899180
Encompass
Change view
Click to view available options
Product Type:
Label Gun
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22899180 Label Gun
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22899180 Supplier Dynamic Diagnostics Supplier No. 310050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quickly label dates on chemicals, reagents, and controls

  • Displays important information with three lines of print and 36 positions
  • Print window clearly displays codes and dates settings for easy adjustment
  • Fast load design with snap-in rollers eliminates messy ink
  • Exceptionally durable–repeated drop tests onto concrete floors
  • Eliminates confusion and clerical errors with clear, highly legible print
  • Lightweight and comfortable to use with easy-squeeze trigger

Order Info

Weight with one roll of labels: 20.8 oz. (590g)

Warranty

One year

Specifications

Product Type Label Gun
For Use With (Equipment) Easy squeeze trigger
Weight (Metric) 0.59 kg