missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaMag™-96 Bottom Magnet DFS Item

Magnet pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200 μL) non-skirted and half-skirted and replaces the Dynal MPC-96B.

Supplier:  Invitrogen™ 12332D

Catalog No. 12332D

Only null left
Explore available promotions

Includes: One magnet

Description

Description

Magnet pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200 μL) non-skirted and half-skirted and replaces the Dynal MPC-96B. When used in combination with the DynaMag-96 Side on an automation platform, the two magnets allow for significant volume changes through a workflow.

  • High wash volumes—DynaMag-96 Side
  • Low Elution Volumes—DynaMag-96 Bottom
  • Concentrates Beads at the bottom of each well
  • Features 96 round magnets (one centered at the bottom of each well: to ensure identical sample handling at each position)
  • Allows for concentrating the sample by eluting in tiny volumes (approx. 10μL)

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Magnet
DynaMag™
DynaMag
Contains one magnet. Store at room temperature.
Room Temperature
1 each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.