missing translation for 'onlineSavingsMsg'
Learn More

Dynalon™ Bio-Bin™ Waste Disposal Container Stands

Numéro de catalogue. p-4876351
Change view
Click to view available options
Length (Metric):
17.01 cm
18.03 cm
42.8 cm
Width (Metric):
17.08 cm
18 cm
27.43 cm
Height (Metric):
19.9 cm
28.57 cm
31.29 cm
Shape:
M Stand
U Stand
Z Stand
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Length (Metric) Width (Metric) Height (Metric) Shape
12009803 18.03 cm 27.43 cm 31.29 cm U Stand
12009804 17.01 cm 17.08 cm 19.9 cm Z Stand
12009805 42.8 cm 18 cm 28.57 cm M Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 12009803
Il en reste null

Accessories for Dynalon Bio-Bin Waste Disposal Containers

For use with Dynalon Bio-Bin Waste Disposal Containers (Cat. No. 12—009—800)

Spécifications

Certifications/Compliance UN 3291
Dimensions (L x W x H) 7.1 x 10.8 x 12.32 in. (18.03 x 27.43 x 31.29 cm)
Height (Metric) 31.29 cm
Length (Metric) 18.03 cm
Shape U Stand
Width (Metric) 27.43 cm
Includes 2 x 2 in. biohazard waste labels, four (4) non slip feet
For Use With (Application) U stand holds the 6L Bio-bin at a 30° angle, making it ideal for use in a fume cabinet, biological safety cabinet, other workstation or bench
For Use With (Equipment) 6.0L Bio-Bin Waste Container
Width (English) 10.8 in.
Length (English) 7.1 in.
Height (English) 12.32 in.
Material Stainless Steel
Product Type Waste Container Stand
Afficher plus de résultats Afficher moins de résultats