missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaGreen™ Protein A/G Magnetic Beads
GREENER_CHOICE

Catalog No. p-7228533
Encompass
Change view
Click to view available options
Quantity:
0.5 mL, 20 Reactions
25 mL, 1000 Reactions
3 mL, 120 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80104G 0.5 mL, 20 Reactions
80105G 3 mL, 120 Reactions
80106G 25 mL, 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 80104G Supplier Invitrogen™ Supplier No. 80104G
Only null left

DynaGreen Protein A/G Magnetic Beads are intended for use in immunoprecipitation (IP) applications while also being an environmentally sustainable choice.

DynaGreen Protein A/G Magnetic Beads are intended for use in immunoprecipitation (IP) applications while also being an environmentally sustainable choice. They are microplastic-free submicron magnetic beads that provide reproducible isolation of target proteins in a simple and efficient workflow that can be performed manually or automated on KingFisher™ instruments.

Benefits of DynaGreen Protein A/G Magnetic Beads include:

  • High performance—reproducible IP with high-yield and low non-specific binding
  • Reduced environmental impact—non-microplastic bead core and holistic sustainable product design
  • Flexible—direct and indirect immunoprecipitation helps ensure isolation of non-abundant proteins
  • Automation ready—easy automation for high-throughput needs on the Kingfisher Sample Purification Systems

Simple, gentle, and efficient manual separation

DynaGreen Protein A/G Magnetic Beads are microplastic-free superparamagnetic beads with recombinant protein A and protein G covalently coupled to the surface. They were developed for convenient IP of target proteins when the IP antibody interacts strongly with Protein A/G (see subclass specificity in the manual). The submicron bead size (∼250 nm) provides low sedimentation and a large target capture surface area. The method is a simple bind-incubate-elute protocol with no pre-clearing required. This results in an efficient high-yield isolation of target protein by direct or indirect IP in less than 80 minutes.

The magnetic separation technology used by DynaGreen Protein A/G Magnetic Beads is rapid and gentle, thereby reducing physical stress to the target proteins. Furthermore, the non-porous nature of the beads lowers the risk of antibodies being trapped and becoming inaccessible. This helps to minimize the amount of antibody required to produce accurate, and reproducible results. The yield and purity of the isolated target protein are highly compatible for use in western blotting and mass spectrometry. The product manual below includes a list of required reagents, materials, and details of compatible buffer recipes.

Rapid and reproducible automated workflow

The DynaGreen magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers. The automated protocol can be performed in just 40 minutes with up to 96 samples per run, which helps reduce hands-on time while maintaining the same reproducible high target yield and low non-specific binding compared to the manual protocol.

A certified choice for environmentally sustainable research

Microplastic-free DynaGreen Protein A/G Magnetic Beads are designed for both high performance in immunoprecipitation workflows and lower environmental impact. They provide exceptional purity and high yield of target proteins compared to other magnetic beads in both direct and indirect immunoprecipitation workflows. At the same time, the product boasts an extensive list of third-party ACT-label-certified sustainable product design elements by incorporating principles of green chemistry and green engineering.

Choosing DynaGreen Protein A/G Magnetic Beads can help reduce the environmental impact of research without compromising on results.

Learn more about our DynaGreen magnetic beads and KingFisher Sample Purification Systems:

  • Alternative sustainable products: DynaGreen Protein A (Cat. No. 80102G) and DynaGreen Capture-Select anti-FC (multispecies) (Cat. No. 80107G)
  • DynaGreen magnetic beads sustainability
  • DynaMag magnets for magnetic bead separations
  • KingFisher automation protocols
  • KingFisher Flex instrument video

Specifications

Concentration 20 mg/mL
Description Recombinant Protein A/G covalently bound to DynaGreen magnetic beads
Material Iron Oxide
Content And Storage DynaGreen Protein A/G Magnetic Beads are supplied in phosphate buffer, pH 7.4, with a biodegradable surfactant and a preservative. Store at 2–8°C.
Diameter (Metric) 250 nm
For Use With (Equipment) KingFisher™ Sample Purification Systems, DynaMag™ Magnets
High-throughput Compatibility High-throughput Compatible
Shipping Condition Ambient temperature
Product Line DynaGreen
Quantity 0.5 mL, 20 Reactions
For Use With (Application) Immunoprecipitation
Target Proteins
Type Protein A/G Magnetic Bead
Ligand Type Recombinant Protein A/G
Sufficient For 20 Tests
Color Brown
Purity or Quality Grade For Research Use Only
Certifications/Compliance ISO9001 and ISO13485
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.