missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Protein G and Magnet Starter Pack

Catalog No. 10017D
Encompass
Change view
Click to view available options
Sufficient For:
100 reactions
40 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
10017D 100 reactions
10014D 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10017D Supplier Invitrogen™ Supplier No. 10017D

missing translation for 'validMainframeMsg'

The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience.

Space-saving design with our smallest thermal cycler footprint

  • Flexible modular design saves space on deck
  • Separate control panel module provides flexibility when integrating on the robotic deck
  • Choice of 10 cm,1 meter and 3 meter cabling options

Automated lid and cycler features

  • 384-well block
  • Automated lid is compatible with programming for easy hands-free operation with liquid handling or plate stackers
  • Heated cover slides forward to cover the plate nest,ensuring thermal contact and eliminating evaporation

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Content And Storage The product contains:
• 5 mL Dynabeads Protein G beads
• 1 x DynaMag-2 magnet

Store Dynabeads at 2°C to 8°C.
Shipping Condition Room Temperature
Product Line DynaMag, Dynabeads
Quantity 100 reactions
Type Magnet Starter Pack
Ligand Type Protein G
Sufficient For 100 reactions

For Research Use Only. Not for use in diagnostic procedures.