missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynabeads™ Protein A and Magnet Starter Pack
The Dynabeads Protein A and Magnet Starter Pack combines Dynabeads Protein A magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience.
$1442.00 - $1442.00
Specifications
| Shipping Condition | Room Temperature |
|---|---|
| Product Line | DynaMag, Dynabeads |
| Type | Magnet Starter Pack |
| Ligand Type | Protein A |
Description
Space-saving design with our smallest thermal cycler footprint
- Flexible modular design saves space on deck
- Separate control panel module provides flexibility when integrating on the robotic deck
- Choice of 10 cm,1 meter, and 3 meter cabling options
Automated lid and cycler features
- 384-well block
- Automated lid is compatible with programming for easy hands-free operation with liquid handling or plate stackers
- Heated cover slides forward to cover the plate nest,ensuring thermal contact and eliminating evaporation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
| Room Temperature | |
| DynaMag, Dynabeads | |
| Magnet Starter Pack | |
| Protein A |
For Research Use Only. Not for use in diagnostic procedures.