missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAVX Magnetic Beads
GREENER_CHOICE

Catalog No. 2853522001
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
2853522001 1 mL
2853522005 5 mL
2853522050 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2853522001 Supplier Thermo Scientific™ Supplier No. 2853522001

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAVX Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications.

Dynabeads CaptureSelect AAVX Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. The anti-AAVX affinity ligand demonstrates good binding reactivity towards a large variety of AAV serotypes, including AAV1 to AAV8, AAVrh10, and synthetic serotypes. The magnetic beads enable purification using manual and robotic magnetic separators of AAV serotypes directly from crude feedstock in a single step with high purity and yield.

Features of DynaBeads CaptureSelect AAVX Magnetic Beads include:

  • One-magnetic-step AAV purification from crude material with high purity and yield
  • Compatible with manual and automated workflows
  • High specificity and capacity
  • Easy upscaling to higher volumes
  • Non-animal derived

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a ∼12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Bead matrix: superparamagnetic
  • Bead size: 4.5 μm
  • Ligand: CaptureSelect AAVX affinity ligand
  • Binding capacity: ∼1 x1012 viral particles/mg beads
  • Elution conditions: 50 mM citric acid pH 2.5-3.0
  • Formulation buffer: tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

Specifications

Binding Capacity ∼1 x 1012 viral particles/mg beads
Capacity (Metric) 1 mL
Concentration 30 mg beads/mL slurry, 1 mg contains approximately 1.3 x 107 beads
Formulation Tris-buffered saline with 0.05% Tween-20 and 0.05% Sodium Azide
Matrix Non-porous monodisperse superparamagnetic beads
Content And Storage Store at 2–8°C.
Particle Size 4.5 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 1 mL
For Use With (Application) Magnetic isolation of AAV capsids
Target Multiple AAV serotypes
Ligand Type AAVX Affinity Ligand
Show More Show Less