missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAV9 Magnetic Beads
GREENER_CHOICE

Catalog No. 2853332050
Change view
Click to view available options
Capacity (Metric):
>0.5 x 1011 vp/mg beads
>5 x 1011 vp/mg beads
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
2853332050 >0.5 x 1011 vp/mg beads
2853332001 >5 x 1011 vp/mg beads
2853332005 >0.5 x 1011 vp/mg beads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2853332050 Supplier Thermo Scientific™ Supplier No. 2853332050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAV9 Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of naturally occurring and synthetic adeno-associated virus serotype 9 (AAV9) used for gene therapy applications.

Dynabeads CaptureSelect AAV9 Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of naturally occurring and synthetic adeno-associated virus serotype 9 (AAV9) used for gene therapy applications. The anti-AAV9 affinity ligand demonstrates good binding reactivity towards the AAV9 serotype. The magnetic beads enable purification using manual and robotic magnetic separators of the AAV9 serotype directly from crude feedstock in a single step with high purity and yield.

Features of DynaBeads CaptureSelect AAV9 Magnetic Beads include:

  • One-magnetic-step AAV purification from crude material with high purity and yield
  • Compatible with manual and automated workflows
  • High specificity and capacity
  • Easy upscaling to higher volumes
  • Non-animal derived

The magnetic beads bind with high affinity specifically to the AAV9 serotype. For the isolation of other AAV serotype capsids we recommend DynaBeads CaptureSelect AAVX magnetic beads (Cat. No. 2853522001, 2853522005, 2853522050).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a ∼12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain. The ligand is efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Bead matrix: non-porous monodisperse superparamagnetic beads
  • Bead size: 4.5 μm
  • Ligand: CaptureSelect AAV9 affinity ligand
  • Binding capacity: >5 x 1011 viral particles/mg beads
  • Elution conditions: 50 mM citric acid, pH 2.5–3.0
  • Formulation buffer: tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

Specifications

Binding Capacity >0.5 x 1011 vp/mg beads
Capacity (Metric) >0.5 x 1011 vp/mg beads
Concentration 30 mg beads/mL slurry, 1 mg contains approximately 1.3 x 107 beads
Formulation 4 to 6% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Matrix Non-porous monodisperse superparamagnetic beads
Content And Storage Store at 2°C to 8°C.
Particle Size 4.5 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
For Use With (Application) Magnetic isolation of AAV9 capsids
Target AAV9 (Adeno-associated serotype 9)
Type Magnetic Affinity Bead
Ligand Type CaptureSelect™ AAV9 Affinity Ligand
Sufficient For Sample Preparation
Show More Show Less