Learn More
DWK Life Sciences DURAN™ Screw Cap GL 45 with 2-hose connector
Catalog No. 05719310
Change view
Click to view available options
Thread Size (Metric):
GL 45
Catalog No. Thread Size (Metric)
05719310 GL 45
1 options
Catalog No. 05719310 Supplier DWK Life Sciences Supplier No. 293102807
A resusable connector GL 45 cap with two ports for connecting tubing.
GL 45 cap connection system for the easy connection of flexible tubing to the DURAN™ GL 45 bottles. The two polyproplylene connectors have angled top and straight underside connectors. Grey polypropylene cap center rotates freely, allowing the bottle to be unscrewed without the need to disconnect the tubing. The cap is ideal for use with soft elastic tubing that has an inner diameter of 6 – 9 mm silicone tubing. An optional venting connector is available with, or without a syringe filter. Highly versatile as the screw cap is based on the standard GL 45 thread. Temperature resistant up to +140°C. Fully autoclavable and washer-proof. Typical applications: Possible biotech applications include the transfer of sterile media from one container to another using a peristaltic pump.
  • Highly versatile as the screw cap is based on the standard GL 45 thread
  • 2-port hose connectors allow easy connection of silicone tubing to lab bottles for liquid transfer
  • EPDM ensures an airtight seal with bottle
  • Grey polypropylene cap center rotates freely, allowing the bottle to be unscrewed without the need to disconnect the tubing
  • Temperature resistant up to +140°C
  • Fully autoclavable / dishwasher-proof
Specifications
Description DURAN Screw Cap GL 45 with 2 hose connections EDPM Gasket
For Use With (Application) Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer bewteen lab bottles; Supply of reagents to instruments
Material Polypropylene, EPDM
Quantity 2/Pk.
Thread Size (Metric) GL 45
Neck Finish 93 mm
Autoclavable Autoclavable