missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Pressure Plus + Laboratory Bottle with Protect Plastic Coating

Safer option for laboratory applications involving vacuum or pressure. Wheaton™ DURAN™ Pressure Plus + Laboratory Bottle with Protect Plastic Coating, have guaranteed pressure resistance and offer optimal safety for user and contents.

$1,579.71 - $2,843.44

Specifications

Material Borosilicate Glass
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
14209170
View Documents Promotion Details
DWK Life Sciences
DWK218152402
100 mL
Case of 10 Each for $2,843.44
Only null left
 
14209174
View Documents Promotion Details
DWK Life Sciences
DWK1175925
250 mL
Case of 10 Each for $1,579.71
Only null left
 
14209175
View Documents Promotion Details
DWK Life Sciences
DWK1175926
500 mL
Case of 10 Each for $1,988.26
Only null left
 
14209176
View Documents Promotion Details
DWK Life Sciences
DWK218155408
1000 mL
Case of 10 Each for $2,626.38
Only null left
 
Description

Description

  • Manufactured from DURAN borosilicate 3.3 glass
  • Vacuum resistance upto I bar vacuum
  • Pressure resistance to + 1.5 bar gauge
  • Certified by the German Technical Inspection Association (TÜV) in accordance with DIN EN 1595:1997-05
  • Features industry standard GL?45 DIN thread
  • Fully autoclavable and sterilizable
  • Protect coated version offers protection for user and contents in the event of breakage due to impact
  • Special geometry still conforms to ISO 4796-1:2016 to maintain instrument compatibility
  • Manufacturing traceability is offered through a lot specific Retrace Code printed on bottle
  • Available in four convenient sizes: 100, 250, 500 and 1000mL
Specifications

Specifications

Borosilicate Glass
Videos
SDS
Documents

Documents