missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GL 45 Hydra Square Bottle
Description
- Ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents
- Five GL 45 necks on the purging bottle increase safety and reduce the risk of cross-contamination and air intake
- Capped bottles reduce the risk of volatile organic compound (VOC) escape and spillage
- GL 45 threads are compatible with standard HPLC connection caps
- Manufactured from borosilicate 3.3 glass, offering a very low inorganic ion extractables profile
- DURAN™ borosilicate glass 3.3 meets ISO 3585 and is classified as Type I, Class A low-expansion borosilicate glass per ASTM-E438-1992
- Space-saving square bottle shape
- Durable white enamel volume graduations
- Retrace Code and certificate support manufacturing lot traceability
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Cleaning Procedure | Lab washer compatible |
| Color | Clear |
| Diameter (Metric) Inner Neck | 30 mm |
| For Use With (Application) | HPLC Purge Bottle, Reagent Supply to Liquid Handling Systems and Analyzers |
| Includes | 4 GL 45 Caps, 4 GL 45 Side Necks |
| Material | Borosilicate Glass (3.3) |
| Length (Metric) | 222.5 mm |
| Max. Temperature (Metric) | 500°C |
| Quantity | 1/Ea. |
| Show More |