Learn More
DuPont™ Tychem™ 4000 Coverall (all)
DuPont Tychem 4000 Coverall. Collar. Elastic Wrists. Attached Socks. Storm Flap with Adhesive Closure. Taped Seams. White.
$202.60 - $265.57
Specifications
Gender Unisex
Color White
Ankle Style Attached Socks
Neck Style Collared
Seam Type Taped
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19813437
View Documents
DuPont™
SL121TWH2X000400
2X-Large
Case of 4 Each for $214.76
Only null left
19813433
View Documents
DuPont™
SL121TWHLG000400
Large
Case of 4 for $202.60
Only null left
19-162-766
View Documents
DuPont™
SL121TWH5X000400
5X-Large
Case of 4 Each for $265.57
Only null left
19-162-765
View Documents
DuPont™
SL121TWH4X000400
4X-Large
Case of 4 Each for $245.87
Only null left
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Specifications
Unisex
Attached Socks
Taped
White
Collared