missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Coverall (all)

Catalog No. 19813437
Change view
Click to view available options
Size:
Large
2X-Large
4X-Large
5X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19813437 2X-Large
19813433 Large
19162766 5X-Large
19162765 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19813437 Supplier DuPont™ Supplier No. SL121TWH2X000400

DuPont Tychem 4000 Coverall. Collar. Elastic Wrists. Attached Socks. Storm Flap with Adhesive Closure. Taped Seams. White.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Taped
Coating Saranex 23-P film
Size (English) Chest 44.25 to 47.75 in.
Size (English) Inseam 30 in.
Size (Metric) Chest 112.39 to 121.28 cm
Size (Metric) Inseam 76.2 cm
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 38.25 in.
Length (Metric) Sleeve 97.15 cm
Width (English) Chest 28.75 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Show More Show Less