Learn More
DuPont™ Tychem™ 4000 Coverall (all)
DuPont Tychem 4000 Coverall. Collar. Elastic Wrists. Attached Socks. Storm Flap with Adhesive Closure. Taped Seams. White.
Supplier:  DuPont™ SL121TWHLG000400
 View more versions of this product
Catalog No. 19813433
Only null left
Explore available promotions
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Specifications
Large
Tyvek
Attached Socks
Collared
Taped
38.25 to 41.75 in.
97.15 to 106.04 cm
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
88.9 cm
5.5 to 5.9 ft.
Unisex
White
Elastic
Zipper
Saranex 23-P film
28.5 in.
72.39 cm
35 in.
25.75 in.
1.67 to 1.8 m
Product Suggestions
Product Suggestions