Learn More
DuPont™ Tychem™ 4000 Coverall (all)
DuPont Tychem 4000 Coverall. Collar. Elastic Wrists. Attached Socks. Storm Flap with Adhesive Closure. Taped Seams. White.
Supplier: DuPont™ SL121TWH5X000400
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Laydown collar
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
| 5X-Large | |
| Tychem | |
| Attached Socks | |
| Front Zipper | |
| Chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments |
| Unisex | |
| White | |
| Collared | |
| Taped |