Learn More
Agilent BioTek Dual Reagent Injector
Dual Reagent Injector is a module that includes syringes, reagent bottles and holders, and inlet tubing.
Supplier: Agilent BioTek 8040036
Description
- Accessory required
- Includes 2 syringes, 2 reagent bottles, 2 bottle holders, and inlet tubing
Identifiants chimiques
| Dual Reagent Injector |
Spécifications
| Dual Reagent Injector | |
| LFX, Cytation |