Learn More
Hoefer Inc Hoefer™ Dual-Gel Caster for SE250&SE260 Mighty Small Mini Vertical Electrophoresis Unit
Numéro de catalogue. NC2741125
Encompass
Enable several gradient or homogenous gels to be cast simultaneouslyDual-Gel Caster:Cast one or two gelsAccommodates both 10 x 8 cm and 10 x 10.5 cm platesUses the same rugged and effective clamp/cam casting method as used with the SE600 and SE400 Series castersThe Dual-Gel Caster includes:Casting CradleCasting Clamp Assemblies, 2 pcsSealing Gasket SetCams, 4 pcsOrder glass plates, SE511 combs and sets of spacers separately.
Numéro de catalogue. NC2741125 Fournisseur Hoefer Inc Code fournisseur SE245
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Enable several gradient or homogenous gels to be cast simultaneouslyDual-Gel Caster:Cast one or two gelsAccommodates both 10 x 8 cm and 10 x 10.5 cm platesUses the same rugged and effective clamp/cam casting method as used with the SE600 and SE400 Series castersThe Dual-Gel Caster includes:Casting CradleCasting Clamp Assemblies, 2 pcsSealing Gasket SetCams, 4 pcsOrder glass plates, SE511 combs and sets of spacers separately.