Learn More
Applied Biosystems™ DS-33 Matrix Standard Kit (Dye Set G5)
Description
Includes
Contains 5 dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution
The Matrix Standard Kit is used to generate the multicomponent matrix required when analyzing 6-FAM, VIC, NED, PET, and LIZ-labeled DNA fragments on the Applied Biosystems SeqStudio, 3500, and 3730 series Systems. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples in a single capillary.
Order Info
Each
Specifications
Specifications
| Shipping Condition | Wet Ice |
| For Use With (Equipment) | 310 Genetic Analyzer, 3100 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer; SeqStudio |
| Quantity | 8 Runs |
Frequently Asked Questions (FAQs)
There is no fixed time as to when a spectral must be run. However, some users prefer to run a new spectral at a preset time to ensure it is up to date. Data quality will be the largest indicator of when a new spectral is required. A separate spectral calibration needs to be performed for each dye set, array type, and array length. For G5-RCT dye set users only: a spectral calibration must be re-run if the array detection cell is moved/repositioned. Refer to FAQ "When should I run a spectral calibration?" for more information on when a spectral should be run.
A new spectral calibration should be run:
- Whenever a new dye set is used on an instrument.
- After any optics adjustment performed by a service engineer.
- If the instrument has been moved.
- If you see a decrease in spectral separation (pull-up and/or pull-down peaks).
- If you alter any conditions (dye set, array type, or array length).
- For G5-RCT, Any4dye-HDR, and S dye set users only: Any time the array detection cell is moved/repositioned.
A spectral calibration is an algorithm applied to raw data, which converts it into the component 4 or 5 dye data stored in the sample files. A spectral is created for a specific dye set (combination of dyes), array type (4 or 16 capillaries), and array length (36cm or 50cm). It is used to correct for the natural overlap of the fluorescent dyes.
DS-02 (Dye Set E5), DS-30 (Dye Set D), DS-31 (Dye Set D), DS-32 (Dye Set F), and DS-33 (Dye Set G5) are all supported on the Applied Biosystems 3130 Series systems. Please refer to the Applied Biosystems 3130/3130xl Genetic Analyzers Getting Started Guide (http://tools.thermofisher.com/content/sfs/manuals/cms_041468.pdf) for more information.
If the peaks in other colors are directly below the peak of interest, the issue could be that the fluorescent dye being used is not part of the selected dye set, the spectral calibration needs to be performed, or the peaks are offscale. Confirm that the dye set selected on the instrument is compatible with the dye being used, run a new spectral calibration if the correct dye set has been selected and, if the signal intensity is too high, decrease sample concentration during PCR or when preparing samples for electrophoresis.
For Research Use Only. Not for use in diagnostic procedures.